P3534
Pentafluorobenzyl bromide
Synonym(s):
α-Bromo-2,3,4,5,6-pentafluorotoluene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
UNSPSC Code:
12352200
Recommended Products
storage temp.
2-8°C
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
181.4 °F - closed cup
Flash Point(C)
83 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wei Liu et al.
Free radical biology & medicine, 47(8), 1101-1107 (2009-08-04)
Free radical-induced lipid peroxidation has been implicated in a number of human diseases including atherosclerosis, cancer, and neurodegenerative diseases. F(2)-Isoprostanes (IsoPs) are isomers of prostaglandin PGF(2alpha) that are generated in vivo from the free radical-initiated peroxidation of arachidonic acid independent
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service