Skip to Content
Merck
All Photos(1)

Documents

P3034

Sigma-Aldrich

Pituitary acetone powder from carp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

biological source

carp

form

powder

storage temp.

−20°C

Application

Pituitary acetone powder from carp has been used to study its effects on Acipenser gueldenstaedtii sperm characteristics. It has also been used to study its effects on the ionic composition, osmolality, and motility of spermatozoa in Polyodon spathula seminal fluid.

Biochem/physiol Actions

Pituitary acetone powder from carp is used commonly by fish farmers to induce ovulation in fishes and carps.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of luteinizing hormone-releasing hormone analogue and carp pituitary hormones on Russian sturgeon (Acipenser gueldenstaedtii) sperm characteristic
Aquaculture Research, 49(2), 1127-1130 (2018)
Ionic composition and osmolality of paddlefish (Polyodon spathula, Acipenseriformes) seminal fluid
Aquaculture International, 11(4), 357-368 (2003)
Comparison of the Efficiency of Common Carp and Silver Carp Pituitary in the Breeding of Common Carp (Cyprinus carpio) and Northern Pike (Esox lucius)
Turkish Journal of Fisheries and Aquatic Sciences, 14, 841-844 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service