Skip to Content
Merck
All Photos(1)

Key Documents

P2742

Sigma-Aldrich

PD 404,182

≥98% (HPLC)

Synonym(s):

6H-6-Imino-(2,3,4,5-tetrahydropyrimido)[1,2-c]-[1,3]benzothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N3S
CAS Number:
Molecular Weight:
217.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: soluble ≥22 mg/mL
H2O: insoluble

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

N=C1Sc2ccccc2C3=NCCCN13

InChI

1S/C11H11N3S/c12-11-14-7-3-6-13-10(14)8-4-1-2-5-9(8)15-11/h1-2,4-5,12H,3,6-7H2

InChI key

JNENSSREQFBZGT-UHFFFAOYSA-N

Biochem/physiol Actions

Potential antibiotic of gram negative bacteria. KDO-8-P synthase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service