Skip to Content
Merck
All Photos(1)

Key Documents

P204

Sigma-Aldrich

Phenylbenzene ω-phosphono-α-amino acid

solid

Synonym(s):

3-[2′-Phosphonomethyl[1,1′-biphenyl]-3-yl]alanine, PMBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18NO5P
Molecular Weight:
335.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

H2O: 0.15 mg/mL
DMSO: 0.2 mg/mL
0.1 M NaOH: >10 mg/mL

SMILES string

N[C@@H](Cc1cccc(c1)-c2ccccc2CP(O)(O)=O)C(O)=O

InChI

1S/C16H18NO5P/c17-15(16(18)19)9-11-4-3-6-12(8-11)14-7-2-1-5-13(14)10-23(20,21)22/h1-8,15H,9-10,17H2,(H,18,19)(H2,20,21,22)/t15-/m0/s1

InChI key

NCEGJIHRQBRVJQ-HNNXBMFYSA-N

Gene Information

human ... GRIN1(2902)

Biochem/physiol Actions

Selective strychnine-sensitive glycine receptor antagonist.

Legal Information

Made and sold under license from Nippon Chemiphar.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Saitoh et al.
British journal of pharmacology, 113(1), 165-170 (1994-09-01)
1. 3-[2'-Phosphonomethyl[1,1'-biphenyl]-3-yl]alanine (PMBA) is a novel glycine antagonist at strychnine-sensitive receptors. The chemical structure of PMBA, possessing both a glycine moiety and a phosphono group, is quite different from that of strychnine. 2. In the spinal motoneurone of newborn rats
K Watanabe et al.
Soft matter, 14(17), 3372-3377 (2018-04-06)
An external electric AC field with a field strength ranging from 10 V mm-1 to 30 V mm-1 and a frequency ranging from 0.1 kHz to 1 MHz was applied to suspensions of gold nanoparticles (Au NPs) to control their
Akihisa Otaka et al.
Journal of biomedical materials research. Part A, 108(10), 2090-2099 (2020-04-24)
Current chemotherapy methods have limited effectiveness in eliminating bone metastasis, which leads to a poor prognosis associated with severe bone disorders. To provide regional chemotherapy for this metastatic tumor, a bone-targeting drug carrier was produced by introducing the osteotropic bisphosphonate
Kanako Watanabe et al.
Journal of colloid and interface science, 566, 202-210 (2020-02-02)
Assembly of plasmonic nanoparticle clusters having hotspots in a specific space is an effective way to efficiently utilize their plasmonic properties. In the assembly, however, bulk-like aggregates of the nanoparticles are readily formed by strong van der Waals forces, inducing
Shahin Aghamiri et al.
IET nanobiotechnology, 13(8), 808-815 (2019-10-19)
Breast cancer is the second cause of death in the world. Ionising radiation is a potent mutagen that can cause DNA damage, chromosomes breakage, and cell death. In the present study, radiotherapy and nanoparticle-antibodies (ABs) have been combined to enhance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service