Skip to Content
Merck
All Photos(2)

Key Documents

P1870

Sigma-Aldrich

Anti-p120ctn antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-p120cas

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 120 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: 1:500 using human epidermoid carcinoma A431 cell line.
microarray: suitable
western blot: 1:2,000 using rat brain extract and human endothelial ECV304 whole cell extract.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNND1(1500)

General description

Catenin delta 1 (CTNND1), the 120 kDa catenin-related protein is also referred as catenin p120ctn, p120cas and pp120 src substrate. p120ctn contains a central armadillo repeat domain (ARM domain) and is structurally similar to β-catenin and γ-catenin. It codes for a member of the Armadillo (Arm) family of proteins. This gene is located on human chromosome 11q12.

Specificity

The sequence is identical in mouse p120ctn and isoforms 1, 2, 3, and 4 of human p120ctn.

Immunogen

synthetic peptide corresponding to the sequence located near the C-terminus of human p120ctn (catenin-related, isoform 1B, amino acids 881-900) conjugated to KLH.

Application

Anti-p120ctn antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Catenin delta 1 (CTNND1/p120ctn) is described as a prominent substrate for the Src oncoprotein and a variety of receptor tyrosine kinases (RTKs) including those for epidermal growth factor (EGF), platelet derived growth factor (PDGF) and colony stimulating factor-1 (CSF-1). It can bind directly to the cytoplasmic domain of the epithelial E-cadherin via the armadillo repeat domain (ARM) domain. It co-precipitates in multiprotein complexes containing E-cadherin or other cadherins such as neural (N)- or placental (P)- cadherins, depending on the cell type. CTNND1 participates in the oncogenesis of some cancer. It can serve as a tumor oncogene in hepatocellular carcinoma (HCC).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For prolonged storage, freeze in working aliquots at −20 °C. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Werner W Franke et al.
Cell and tissue research, 379(1), 75-92 (2019-11-13)
In the molecular biological and ultrastructural studies of the peritubular wall cells encasing the seminiferous tubules of mammalian testes, we found it necessary to characterize the outermost cell layer bordering on the interstitial space in detail. For half a century
EXPRESSION AND CLINICAL SIGNIFICANCE OF P120 CATENIN M RNA AND PROTEIN IN PANCREATIC CARCINOMA
Fei Y, et al.
Bosnian Journal of Basic Medical Sciences / Udruzenje Basicnih Mediciniskih Znanosti = Association of Basic Medical Sciences, 9(3), 191-191 (2009)
The catenin p120 ctn interacts with Kaiso, a novel BTB/POZ domain zinc finger transcription factor
Daniel JM and Reynolds AB
Molecular and Cellular Biology, 19(5), 3614-3623 (1999)
Overexpression of CTNND1 in hepatocellular carcinoma promotes carcinous characters through activation of Wnt/beta-catenin signaling
Tang B, et al.
Journal of Experimental & Clinical Cancer Research, 35(1), 82-82 (2016)
Structure of the armadillo repeat domain of plakophilin 1
Choi HJ and Weis WI
Journal of Molecular Biology, 346(1), 367-376 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service