Skip to Content
Merck
All Photos(1)

Key Documents

P126

Sigma-Aldrich

Pirenperone

>97%, solid

Synonym(s):

3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2-methyl-4H-pyrido-[1,2-a] pyrimidin-4-one, R-47,465

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24FN3O2
CAS Number:
Molecular Weight:
393.45
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

>97%

form

solid

color

light yellow

solubility

0.1 M HCl: 11 mg/mL
methanol: 2.5 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 5.0 mg/mL

SMILES string

CC1=C(CCN2CCC(CC2)C(=O)c3ccc(F)cc3)C(=O)N4C=CC=CC4=N1

InChI

1S/C23H24FN3O2/c1-16-20(23(29)27-12-3-2-4-21(27)25-16)11-15-26-13-9-18(10-14-26)22(28)17-5-7-19(24)8-6-17/h2-8,12,18H,9-11,13-15H2,1H3

InChI key

HXCNRYXBZNHDNE-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

5-HT2 serotonin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Griebel et al.
Pharmacology, biochemistry, and behavior, 51(2-3), 235-244 (1995-06-01)
The mouse defense test battery (MDTB) has been designed to investigate defensive reactions in Swiss-Webster mice to situations associated with a natural predator, the rat, such as flight, avoidance, defensive threat, defensive attack, and risk assessment activities. The present study
C Goodrich et al.
Physiology & behavior, 46(2), 203-209 (1989-08-01)
Experiments were designed to distinguish between central and peripheral effects on temperature preference and body temperature of drugs injected intraperitoneally (IP) in infant mice ranging in age from 3 to 10 days postpartum. These compared a drug restricted to the
J C Winter et al.
Pharmacology, biochemistry, and behavior, 62(3), 543-547 (1999-03-18)
Stimulus control was established in a group of nine rats using a dose of EGb 761 of 10 mg/kg, administered i.p., 15 min before training. A two-lever operant task using a fixed-ratio 10 schedule of sweetened milk reinforcement was used.
H J Altman et al.
Pharmacology, biochemistry, and behavior, 28(3), 353-359 (1987-11-01)
The experiments examined the effects of acute administration of three different serotonergic receptor antagonists (ketanserin, pirenperone and mianserin) on one-trial passive avoidance retention in mice. Administration of each antagonist 30 min before training produced a dose-dependent impairment in retention. In
L A Dykstra et al.
Psychopharmacology, 112(1), 116-120 (1993-01-01)
The kappa opioid, U50,488, was examined alone and in combination with the 5HT2 antagonists, ketanserin, pirenperone and LY 53857. Squirrel monkeys responded under a shock titration procedure in which shock intensity increased every 15 s from 0.01 to 2.0 mA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service