Skip to Content
Merck
All Photos(1)

Key Documents

P1176

Sigma-Aldrich

Hexaethylene glycol monooctadecyl ether

Synonym(s):

C18E6, Octadecylhexaglycol, Polyoxyethylene (6) stearyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17(OCH2CH2)6OH
CAS Number:
Molecular Weight:
534.81
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

SMILES string

CCCCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service