Skip to Content
Merck
All Photos(1)

Documents

P1076

Sigma-Aldrich

Palmitic acid-[9,10-3H]

ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H32O2
CAS Number:
Molecular Weight:
256.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

ethanol solution

extent of labeling

30-60 Ci/mmol

mass shift

M+4

shipped in

dry ice

storage temp.

−20°C

SMILES string

[3H]C(CCCCCC)C([3H])CCCCCCCC(O)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kumar Chokalingam et al.
Tissue engineering. Part A, 15(9), 2561-2570 (2009-02-05)
The objectives of this study were to determine how tensile stimulation delivered up to 14 days in culture influenced type I collagen gene expression in stem cells cultured in collagen sponges, and to establish if gene expression, measured using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service