OGS1498
PSF-CMV-PURO-NH2-GAUS - GAUSSIA (LUCIFERASE) SECRETION PLASMID
plasmid vector for molecular cloning
Synonym(s):
cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector
About This Item
form
buffered aqueous solution
mol wt
size 6194 bp
bacteria selection
kanamycin
mammalian cells selection
puromycin
Origin of replication
pUC (500 copies)
Peptide cleavage
no cleavage
Peptide tag location
N-terminal
Promoter
Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian
reporter gene
none
Secretion signal
gaussia
shipped in
ambient
storage temp.
−20°C
General description
About the Peptide Tag:This plasmid vector contains an Gaussia luciferase (Gaus) secretory signal peptide (SP) to allow proteins to be exported from the cytosol. During translocation from the cytosol the signal peptide is removed from the protein by endogenous proteases.
Promoter Expression Level: This molecular cloning vector contains the mammalian CMV promoter to drive gene expression. We have tested all of our mammalian promoters in a range of cell types and CMV is consistently the strongest in those we have studied. However there are many reports of the CMV promoter demonstrating silencing by methylation in long-term culture.
Sequence
Genebank Vector Sequence File
FASTA Vector Sequence File
Full Plasmid Map
Analysis Note
Other Notes
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service