Skip to Content
Merck
All Photos(1)

Key Documents

O5508

Sigma-Aldrich

N4-Octadecylcytosine β-D-arabinofuranoside

Synonym(s):

N(4)-octadecyl-ara-C, N4-Stearyl-Ara C, NOAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H49N3O5
CAS Number:
Molecular Weight:
495.70
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

SMILES string

CCCCCCCCCCCCCCCCCCNC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O

InChI

1S/C27H49N3O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-28-23-18-20-30(27(34)29-23)26-25(33)24(32)22(21-31)35-26/h18,20,22,24-26,31-33H,2-17,19,21H2,1H3,(H,28,29,34)/t22-,24-,25+,26-/m1/s1

InChI key

HQHQCEKUGWOYPS-URBBEOKESA-N

Application

N4-Stearyl-Ara C (N(4)-octadecyl-ara-C, NOAC) is a lipophilic derivative of Ara-C used to improve the incorporation of Ara-C, an anti-cancer agent, into lipid bilayers of liposomes and/or target cells.

Other Notes

Lipophilic, cytotoxic derivative of cytosine arabinoside

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K M Rentsch et al.
Journal of chromatography. B, Biomedical applications, 673(2), 259-266 (1995-11-17)
N4-Hexadecyl- and N4-octadecyl-1-beta-D-arabinofuranosylcytosine (NHAC, NOAC) are two new cytostatic derivatives of cytosine arabinoside (ara-C) with improved cytostatic activity and stability against deamination. A high-performance liquid chromatography (HPLC) method was developed for the specific determination of NHAC and NOAC in plasma
Novel PEG-matrix metalloproteinase-2 cleavable peptide-lipid containing galactosylated liposomes for hepatocellular carcinoma-selective targeting.
Terada T, Iwai M, Kawakami S, et al.
J. Controlled Release, 111, 333-342 (2006)
S K Koller-Lucae et al.
British journal of cancer, 80(10), 1542-1549 (1999-07-17)
Low density lipoprotein (LDL) receptor-mediated uptake and cytotoxic effects of N4-octadecyl-1-beta-D-arabinofuranosylcytosine (NOAC) were studied in Daudi lymphoma cells. NOAC was either incorporated into LDL or liposomes to compare specific and unspecific uptake mechanisms. Binding of LDL to Daudi cells was
S K Koller-Lucae et al.
Drug metabolism and disposition: the biological fate of chemicals, 27(3), 342-350 (1999-03-04)
Metabolism and excretion of the new antitumor drug N4-octadecyl-1-beta-D-arabinofuranosylcytosine (NOAC) was investigated in mice. Mice were injected i.v. with tritium-labeled liposomal NOAC (4 micromol/mouse). Analysis of HPLC-purified extracts of liver homogenates by liquid chromatography coupled with mass spectrometry revealed only
Reto Schwendener et al.
Methods in enzymology, 391, 58-70 (2005-02-22)
Highly lipophilic drugs can be used therapeutically only by the addition of possibly toxic solubilizing agents or by development of complex pharmaceutical formulations. One way of overcoming these disadvantages is the incorporation of such drugs into the bilayer matrix of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service