vapor density
1 (vs air)
form
liquid
refractive index
n20/D 1.396 (lit.)
bp
175-176 °C (lit.)
mp
17-18 °C (lit.)
density
0.956 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
InChI
1S/C8H24O4Si4/c1-13(2)9-14(3,4)11-16(7,8)12-15(5,6)10-13/h1-8H3
InChI key
HMMGMWAXVFQUOA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Repr. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
123.8 °F - closed cup
Flash Point(C)
51 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service