Skip to Content
Merck
All Photos(1)

Key Documents

N9879

Sigma-Aldrich

NADase from porcine brain

≥0.007 unit/mg solid

Synonym(s):

DPN Nucleosidase, NAD+ glycohydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€290.00

€290.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€290.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

€290.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

Quality Level

specific activity

≥0.007 unit/mg solid

solubility

insoluble

storage temp.

−20°C

General description

NADase is a glycohydrolase that catalyzes ADP-ribose transfer. [1]

Application

NADase from porcine brain has been used in a study to investigate histidine and related compounds resulting from catalyzed ADP-riboslyation. [2] It has also been used in a study to investigate the preparation of arylazide-substituted pyridine adenine dinucleotides for photoaffinity labeling. [3]

Biochem/physiol Actions

Nicotinamide has been shown to inhibit NADase activity. [4]

Unit Definition

One unit will hydrolyze 1.0 μmole of β-NAD to nicotinamide and ADP-ribose per min at pH 7.3 at 37 °C.

Physical form

Acetone-dried powder

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ding-bao Chen et al.
Zhonghua bing li xue za zhi = Chinese journal of pathology, 41(9), 607-612 (2012-11-20)
To study the clinical and histopathologic features, diagnosis, pathogenesis and therapy of post-transplant lymphoproliferative disorders (PTLD). The clinical and pathologic features of 15 cases of PTLD were retrospectively analyzed by light microscopy, immunohistochemistry and in-situ hybridization, according to the updated
Nicholas Chiorazzi
Hematology. American Society of Hematology. Education Program, 2012, 76-87 (2012-12-13)
Several prognostic markers based on genetic, phenotypic, and molecular characteristics of chronic lymphocytic leukemia (CLL) B cells have emerged in the past decade. The clinical utility of these newer prognostic indicators, alone or in combination with each other and other
Shoko Nakayama et al.
Hematology (Amsterdam, Netherlands), 17(6), 317-320 (2012-11-22)
The accurate determination of cytoplasmic immunoglobulin (cIg) light chain (LC) expression is important to differentiate reactive plasmacytosis from a clonal plasma cell neoplasm such as plasma cell myeloma (PCM). Through retrospective analysis, we studied the cytoplasmic kappa/lambda ratio of CD38-positive
K A Wall et al.
The Biochemical journal, 335 ( Pt 3), 631-636 (1998-10-31)
Carba-NAD and pseudocarba-NAD are carbocyclic analogues of NAD+ in which a 2,3-dihydroxycyclopentane methanol replaces the beta-d-ribonucleotide ring of the nicotinamide riboside moiety of NAD+ [Slama and Simmons (1988) Biochemistry 27, 183-193]. These carbocyclic NAD+ analogues, related to each other as
Julien Calvo et al.
PloS one, 7(11), e50495-e50495 (2012-12-05)
Hematopoietic stem cells are responsible for the generation of the entire blood system through life. This characteristic relies on their ability to self renew and on their multi-potentiality. Thus quantification of the number of hematopoietic stem cells in a given

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service