Skip to Content
Merck
All Photos(1)

Key Documents

N7287

Sigma-Aldrich

NU-1025

≥97% (HPLC), solid

Synonym(s):

8-Hydroxy-2-methylquinazolin-4[3H]-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€199.00

€199.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€199.00

About This Item

Empirical Formula (Hill Notation):
C9H8N2O2
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€199.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

solid

color

off-white

solubility

DMSO: soluble >20 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

CC1=Nc2c(O)cccc2C(=O)N1

InChI

1S/C9H8N2O2/c1-5-10-8-6(9(13)11-5)3-2-4-7(8)12/h2-4,12H,1H3,(H,10,11,13)

InChI key

YJDAOHJWLUNFLX-UHFFFAOYSA-N

Gene Information

mouse ... Parp2(11546)

Biochem/physiol Actions

NU-1025 is a PARP inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evangelos Koustas et al.
Journal of cellular and molecular medicine, 24(18), 10420-10431 (2020-07-21)
Gastric cancer is the fifth most common malignancy and the third leading cause of cancer-related death worldwide. Activation of c-MET increases tumour cell survival through the initiation of the DNA damage repair pathway. PARP is an essential key in the
David Zada et al.
Molecular cell, 81(24), 4979-4993 (2021-11-20)
The characteristics of the sleep drivers and the mechanisms through which sleep relieves the cellular homeostatic pressure are unclear. In flies, zebrafish, mice, and humans, DNA damage levels increase during wakefulness and decrease during sleep. Here, we show that 6 h
Quanju Zhao et al.
Journal of immunology (Baltimore, Md. : 1950), 198(7), 2935-2942 (2017-02-22)
IL-12 and IL-23 are important host defense factors produced by APCs against certain intracellular and extracellular pathogens. Their dysregulation has also been implicated in several autoimmune diseases. The nucleotide polymorphism in the promoter region of Il12b (rs41292470 consisting of the
Michael M Murata et al.
Molecular biology of the cell, 30(20), 2584-2597 (2019-08-08)
DNA damage signaling is critical for the maintenance of genome integrity and cell fate decision. Poly(ADP-ribose) polymerase 1 (PARP1) is a DNA damage sensor rapidly activated in a damage dose- and complexity-dependent manner playing a critical role in the initial
Yang Sun et al.
American journal of respiratory cell and molecular biology, 51(6), 738-749 (2014-05-31)
Lymphangioleiomyomatosis (LAM) is a female-predominant cystic lung disease that can lead to respiratory failure. LAM cells typically have inactivating tuberous sclerosis complex 2 (TSC2) mutations and mammalian target of rapamycin (mTOR) complex (mTORC) 1 activation. Clinical response to the mTORC1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service