Skip to Content
Merck
All Photos(1)

Key Documents

N6910

Sigma-Aldrich

Neomycin trisulfate salt hydrate

Biotechnology Performance Certified, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46N6O13 · 3H2SO4 · xH2O
CAS Number:
Molecular Weight:
908.88 (anhydrous basis)
EC Number:
UNSPSC Code:
51101500
PubChem Substance ID:

grade

Biotechnology Performance Certified

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

SMILES string

O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.NC[C@@H]1O[C@H](O[C@H]2[C@@H](O)[C@@H](O[C@@H]2CO)O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)[C@H]3O[C@H]4O[C@H](CN)[C@@H](O)[C@H](O)[C@H]4N)C(N)[C@@H](O)[C@@H]1O

InChI

1S/C23H46N6O13.3H2O4S.H2O/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;3*1-5(2,3)4;/h5-23,30-36H,1-4,24-29H2;3*(H2,1,2,3,4);1H2/t5-,6+,7-,8+,9-,10-,11?,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+;;;;/m1..../s1

InChI key

WHAGUNPVKDUVFV-QGTTWHFQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Neomycin Trisulfate is produced by Streptomyces containing a minimum of 85% neomycin B. It has been used as a selection agent for prokaryotic cells transformed using the neo selectable marker gene, and to study ototoxic side effects of antibiotics, platelet-derived growth factor responses in certain fibroblasts, and extraction of Nuclear Phosphatidylinositol 4,5-Bisphosphate-Interacting Proteins †††. It is recommended for use in cell culture applications at 50 mg/L. This product is biotechnology performance certified and cell culture tested.

Biochem/physiol Actions

Mode of action: This product acts by binding to the 30S and 50S subunits, causing miscoding and inhibiting initiation and elongation during protein synthesis. Neomycin also blocks voltage-sensitive Ca2+ channels without affecting the Na+/Ca2+ antiporter in neurons.

Antimicrobial spectrum: Neomycin acts against both gram-positive and gram-negative bacteria.

Caution

Stock solutions should be filter sterilized and stored at 2-8°C, and are stable at 37°C for 5 days. A 1 mg/mL neomycin solution in .1 M phosphate buffer, pH 8.0 should be used within 14 days when stored at 0-5°C. Solutions should be protected from light or moisture.

Preparation Note

Neomycin sulfate is soluble in H2O at 50 mg/mL, yielding a clear solution.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F S Vassbotn et al.
The Journal of biological chemistry, 267(22), 15635-15641 (1992-08-05)
The aminoglycoside neomycin has recently been found to affect certain platelet-derived growth factor (PDGF) responses in C3H/10T1/2 C18 fibroblasts. Using porcine aortic endothelial cells transfected with PDGF alpha- or beta-receptors, we explored the possibility that neomycin interferes with the interaction
Michael G Leitner et al.
Molecular pharmacology, 79(1), 51-60 (2010-10-12)
Aminoglycoside antibiotics (AGs) are severely ototoxic. AGs cause degeneration of outer hair cells (OHCs), leading to profound and irreversible hearing loss. The underlying mechanisms are not fully understood. OHC survival critically depends on a specific K+ conductance (I(K,n)) mediated by
Aurélia E Lewis et al.
Molecular & cellular proteomics : MCP, 10(2), M110-M110 (2010-11-05)
Considerable insight into phosphoinositide-regulated cytoplasmic functions has been gained by identifying phosphoinositide-effector proteins. Phosphoinositide-regulated nuclear functions however are fewer and less clear. To address this, we established a proteomic method based on neomycin extraction of intact nuclei to enrich for
Sung Ryeol Park et al.
Natural product reports, 30(1), 11-20 (2012-11-28)
The 2-deoxystreptamine-containing aminoglycosides, such as neomycin, kanamycin and gentamicin, are an important class of antibiotics. A detailed understanding of the complete biosynthetic pathway of aminoglycosides and their biosynthetic enzymes will allow us to not only generate more robust antibiotic agents
Wenlin Zhou et al.
Biology of reproduction, 87(6), 144-144 (2012-10-27)
The domesticated silkworm Bombyx mori L. has important roles in basic biological research and applied science. To explore the practical use of transgenic technology in agricultural silkworm varieties, we fused the neomycin-resistance gene (Neo(R)) and the green fluorescent protein gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service