Skip to Content
Merck
All Photos(1)

Key Documents

N6664

Sigma-Aldrich

Neurodazine

≥98% (HPLC)

Synonym(s):

2-[5-(3-Chlorophenyl)-2-furanyl]-4,5-bis(4-methoxyphenyl)-1H imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€179.00
25 MG
€439.00

€179.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€179.00
25 MG
€439.00

About This Item

Empirical Formula (Hill Notation):
C27H21ClN2O3
CAS Number:
Molecular Weight:
456.92
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.25

€179.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

COc1ccc(cc1)-c2nc([nH]c2-c3ccc(OC)cc3)-c4ccc(o4)-c5cccc(Cl)c5

InChI

1S/C27H21ClN2O3/c1-31-21-10-6-17(7-11-21)25-26(18-8-12-22(32-2)13-9-18)30-27(29-25)24-15-14-23(33-24)19-4-3-5-20(28)16-19/h3-16H,1-2H3,(H,29,30)

InChI key

FEEOFPAEDSMOTO-UHFFFAOYSA-N

Biochem/physiol Actions

Neurodazine is a promoter of neurogenesis. It induces a generation of new nerve cells from skeletal muscle fibers both immature, differentiated myotubes and mature skeletal muscle. Neurodazine promotes the expression of neuron-specific markers in treated C2C12 cells (neurogenesis). In addition, in conjunction with a microtubule-destabilizing agent, Neurodazine allows neurogenic conversion of both differentiated immature myotubes and mature skeletal muscle. Neurodazine is a potential alternative approach to using stem cells.
Neurodazine is a promoter of neurogenesis; induces generation of new nerve cells from skeletal muscle fibers both immature differentiated myotubes and mature skeletal muscle; a potential alternative approach to using stem cells.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Ding et al.
Journal of Alzheimer's disease : JAD, 37(3), 587-592 (2013-08-21)
Ciliary neurotrophic factor (CNTF) is a pleiotropic cytokine that has been fully studied for its structure, receptor, and signaling pathways and its multiplex effects on neural system, skeletal muscle, and weight control. Recent research demonstrates that CNTF also plays an
Ronald W Irwin et al.
Frontiers in endocrinology, 2, 117-117 (2011-01-01)
The proliferative pool and regenerative potential of neural stem cells diminishes with age, a phenomenon that may be exacerbated in prodromal and mild Alzheimer's disease (AD) brains. In parallel, the neuroactive progesterone metabolite, allopregnanolone (APα), along with a host of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service