Skip to Content
Merck
All Photos(1)

Key Documents

N6534

Sigma-Aldrich

NFκB Control Peptide

p50, solution, human

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€222.00

€222.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€222.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

€222.00


Please contact Customer Service for Availability

Product Name

NFκB (p50) Control Peptide human, solution

biological source

human

Quality Level

form

solution

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NFKB1(4790)

Amino Acid Sequence

Ala-Glu-Asp-Asp-Pro-Tyr-Leu-Gly-Arg-Pro-Glu-Gln-Met-Phe

Application

For use as a blocking peptide for anti-NFκB (p50) subunit.

Physical form

Solution containing 0.01% sodium azide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service