Skip to Content
Merck
All Photos(1)

Documents

N6409

Sigma-Aldrich

NFκB (c-Rel) Control Peptide

~1 mg/mL, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

form

buffered aqueous solution

Quality Level

concentration

~1 mg/mL

storage temp.

−20°C

Application

For use as a blocking peptide for anti-NFκB(c-Rel) subunit.

Physical form

Solution in 0.02 M potassium phosphate buffer, pH 7.2, containing 0.15 M NaCl and 0.01% sodium azide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service