Skip to Content
Merck
All Photos(1)

Key Documents

N5270

Sigma-Aldrich

6-Norlysergic acid diethylamide

powder, ≥91% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O
CAS Number:
Molecular Weight:
309.41
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥91% (TLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

orange-tan

solubility

alcohol: soluble

application(s)

forensics and toxicology
veterinary

storage temp.

−20°C

SMILES string

[H][C@@]12Cc3c[nH]c4cccc(C1=C[C@H](CN2)C(=O)N(CC)CC)c34

InChI key

SUXLVXOMPKZBOV-CXAGYDPISA-N

Biochem/physiol Actions

LSD analog

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Hoja et al.
Journal of chromatography. B, Biomedical sciences and applications, 692(2), 329-335 (1997-05-09)
A sensitive and highly specific method for the determination of LSD and N-demethyl-LSD in urine, using combined liquid chromatography and mass spectrometry (LC-MS) with electrospray ionization, has been developed. Extrelut-3 extraction cartridges were used for a basic sample clean-up. Elution
J de Kanel et al.
Journal of forensic sciences, 43(3), 622-625 (1998-06-03)
A forensic procedure for the quantitative confirmation of lysergic acid diethylamide (LSD) and the qualitative confirmation of its metabolite, N-demethyl-LSD, in blood, serum, plasma, and urine samples is presented. The Zymark RapidTrace was used to perform fully automated solid-phase extractions
Quantification of LSD and N-demethyl-LSD in urine by gas chromatography/resonance electron capture ionization mass spectrometry.
H K Lim et al.
Analytical chemistry, 60(14), 1420-1425 (1988-07-15)
Gebhard Thoma et al.
Bioorganic & medicinal chemistry letters, 19(21), 6185-6188 (2009-09-29)
The special ergoline 1 is a highly potent, selective antagonist of the chemokine receptor CXCR3. The surprising selectivity of this LSD-related compound can be explained by different electronic and steric properties of the ergoline core structure caused by the urea

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service