N4763
Nigrosin water soluble
powder, BioReagent, suitable for cell culture
Synonym(s):
Acid black 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Colour Index Number:
50420
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
product line
BioReagent
form
powder
technique(s)
cell culture | mammalian: suitable
solubility
H2O: 10 mg/mL
Looking for similar products? Visit Product Comparison Guide
General description
Nigrosin is an acidic dye, useful for negative staining, in which only the background is stained.
Application
Nigrosin water soluble has been used to stain live sperm.
Used as a viable cell staining agent especially in stem cell biology.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Association of chromosomal aberrations and semen quality in egyptian buffalo bulls
Rushdi HE, et al.
Egyptian journal of animal production, 54(2), 95-103 (2017)
Biotechnology-II: Including Cell Biology, Genetics, Microbiology (2007)
Effect of two protocols of cryopreservation on fertilizing capacity of stallion (Equus caballus) semen
Restrepo BG, et al.
Revista Facultad Nacional de Agronomia (Medellin, Colombia), 65(2), 6711-6718 (2012)
Lukas Schertel et al.
Journal of the Royal Society, Interface, 17(166), 20200196-20200196 (2020-05-21)
Vivid colours found in living organisms are often the result of scattering from hierarchical nanostructures, where the interplay between order and disorder in their packing defines visual appearance. In the case of Flavobacterium IR1, the complex arrangement of the cells
M L Foster et al.
Theriogenology, 76(2), 334-341 (2011-04-19)
Sperm membrane integrity (SMI) is thought to be an important measure of stallion sperm quality. The objective was to compare three methods for evaluating SMI: flow cytometry using SYBR-14/propidium iodide (PI) stain; an automated cell counting device using PI stain;
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service