Skip to Content
Merck
All Photos(1)

Key Documents

N4641

Supelco

11-Nor-Δ8-tetrahydrocannabinol-9-carboxylic acid

vacuum-dried powder

Synonym(s):

Δ6-Tetrahydrocannabinol-7-oic acid, 9-Carboxy-11-nor-Δ8-tetrahydrocannabinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

form

vacuum-dried powder

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

storage temp.

−20°C

SMILES string

CCCCCc1cc(O)c2[C@@H]3CC(=CC[C@H]3C(C)(C)Oc2c1)C(O)=O

Application

Standard for identification of THC metabolites.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schwartz, A., et al.
The Journal of Organic Chemistry, 51, 5463-5463 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service