Skip to Content
Merck
All Photos(1)

Key Documents

N3502

Sigma-Aldrich

2-Nitrophenyl stearate

crystalline

Synonym(s):

Octadecanoic acid 2-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NO4
CAS Number:
Molecular Weight:
405.57
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

crystalline

color

off-white

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)Oc1ccccc1N(=O)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service