Skip to Content
Merck
All Photos(1)

Key Documents

N3261

SAFC

Nylidrin hydrochloride

~95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25NO2 · HCl
CAS Number:
Molecular Weight:
335.87
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

~95%

SMILES string

Cl.CC(CCc1ccccc1)NC(C)C(O)c2ccc(O)cc2

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

β-Adrenergic agonist; peripheral vasodilator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Yuan et al.
Nature chemical biology, 5(10), 765-771 (2009-09-08)
Studies of gene function and molecular mechanisms in Plasmodium falciparum are hampered by difficulties in characterizing and measuring phenotypic differences between individual parasites. We screened seven parasite lines for differences in responses to 1,279 bioactive chemicals. Hundreds of compounds were
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service