Skip to Content
Merck
All Photos(1)

Documents

N2786

Sigma-Aldrich

Monoclonal Anti-Nedd8 antibody produced in mouse

clone ND23, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ND23, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

microarray: suitable
western blot: 0.15-0.3 μg/mL using recombinant GST-Nedd8

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEDD8(4738)

General description

Monoclonal Anti-Nedd8 (mouse IgG2b isotype) is derived from the ND23 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to amino acids of human Nedd8 conjugated keyhole limpet hemocyanin (KLH). Nedd8 is a small protein (9 kDa) that shows the highest similarity to the ubiquitin protein (60% amino acid sequence identity).
Neural precursor cell expressed, developmentally down-regulated 8 (NEDD8) is an ubiquitin-like, 76 amino acid protein. It is expressed in endocrine and epithelial cells. NEDD8 belongs to the ubiquitin-like protein superfamily.

Immunogen

synthetic peptide corresponding to amino acids 20-32 of human Nedd8, conjugated to KLH.

Application

Monoclonal Anti-Nedd8 antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry

Biochem/physiol Actions

Nedd8 conjugation has a physiological role in processes where the stability of nuclear factor kappa light chain enhancer of activated B cells (NF-κB) and p27 is important.
Neural precursor cell expressed, developmentally down-regulated 8 (NEDD8) is involved in pathways concerned with protein degradation. It targets substrates for neddylation, which is a post-translational modification. NEDD8 also regulates the signaling of proteins like cullins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MLN4924, an NAE inhibitor, suppresses AKT and mTOR signaling via upregulation of REDD1 in human myeloma cells
Gu Y, et al.
Blood, 123(21), 3269-3276 (2014)
Angela Lucariello et al.
In vivo (Athens, Greece), 27(4), 501-506 (2013-07-03)
Neural precursor cell-expressed, developmentally down-regulated-8 (NEDD8) is a 76-amino-acid ubiquitin-like polypeptide. NEDD8 affects the signaling of various molecules but the major cellular target proteins are cullins. The neddylation process is correlated closely with apoptosis, cell-cycle regulation, embryogenesis and development. The
Marihan Hegazy et al.
Current protocols in cell biology, 89(1), e115-e115 (2020-10-13)
Biochemical methods can reveal stable protein-protein interactions occurring within cells, but the ability to observe transient events and to visualize the subcellular localization of protein-protein interactions in cells and tissues in situ provides important additional information. The Proximity Ligation Assay®
Yanchun Wang et al.
Cancer biology & therapy, 16(3), 420-429 (2015-03-18)
Recent studies indicate that post-translational protein neddylation is required for the maintenance of cell viability in several lymphoma cell lines, while inhibition of the neddylation pathway with an NEDD8-activating enzyme (NAE) inhibitor MLN4924 induces apoptosis in lymphoma cells. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service