Skip to Content
Merck
All Photos(5)

Documents

N1875

Sigma-Aldrich

Naphthol AS-BI β-D-glucuronide

β-glucuronidase substrate, ≥99% (TLC), powder

Synonym(s):

O-β-D-Glucuronosyl-naphthol AS-BI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22BrNO9
CAS Number:
Molecular Weight:
548.34
Beilstein:
6883214
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

Naphthol AS-BI β-D-glucuronide, β-glucuronidase substrate

Quality Level

Assay

≥99% (TLC)

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

COc1ccccc1NC(=O)c2cc3cc(Br)ccc3cc2O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(O)=O

InChI

1S/C24H22BrNO9/c1-33-16-5-3-2-4-15(16)26-22(30)14-9-12-8-13(25)7-6-11(12)10-17(14)34-24-20(29)18(27)19(28)21(35-24)23(31)32/h2-10,18-21,24,27-29H,1H3,(H,26,30)(H,31,32)/t18-,19-,20+,21-,24+/m0/s1

InChI key

ACOOAEDFQKSZTC-NABGWTBKSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Substrate for histochemical demonstration of β-glucuronidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco A Passini et al.
Methods in molecular biology (Clifton, N.J.), 246, 225-236 (2004-02-19)
The efficient transduction of postmitotic cells by adeno-associated virus (AAV) makes it an excellent vector to deliver marker, functional, or therapeutic genes to the mammalian brain. An attractive feature of AAV is that all the viral-coding sequences are removed when
I Marigómez et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 141(2), 188-193 (2005-07-15)
Digestive cell lysosomes in mussels are known to respond to individual organic chemicals and metals after experimental exposure under laboratory conditions but reports dealing with the response to mixtures of pollutants are scarce. The aim of the present investigation was
Hayashi, M, et al.
The Journal of Histochemistry and Cytochemistry, 12, 293-293 (1963)
Lentiviral vectors for gene transfer to the central nervous system. Applications in lysosomal storage disease animal models.
Deborah J Watson et al.
Methods in molecular medicine, 76, 383-403 (2003-01-16)
Steven J Brooks et al.
Aquatic toxicology (Amsterdam, Netherlands), 160, 1-12 (2015-01-09)
Mytilus species are important organisms in marine systems being highly abundant and widely distributed along the coast of Europe and worldwide. They are typically used in biological effects studies and have a suite of biological effects endpoints that are frequently

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service