Skip to Content
Merck
All Photos(3)

Key Documents

N1655

Sigma-Aldrich

N-Z-Amine® YT

Synonym(s):

Peptone from casein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine milk (casein)

form

solid

nitrogen analysis

~6.3% amino, ~13.2% total

Looking for similar products? Visit Product Comparison Guide

Legal Information

N-Z-Amine is a registered trademark of Kerry Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service