Skip to Content
Merck
All Photos(2)

Documents

N158

Sigma-Aldrich

Naftopidil hydrochloride hydrate

solid

Synonym(s):

4-(2-Methoxyphenyl)-α-[(1-naphthalenyloxy)methyl]-1-piperazineethanol hydrochloride hydrate, KT-611

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28N2O3 · xHCl · yH2O
CAS Number:
Molecular Weight:
392.49 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

methanol: >10 mg/mL
H2O: insoluble

SMILES string

O.Cl.COc1ccccc1N2CCN(CC2)CC(O)COc3cccc4ccccc34

InChI

1S/C24H28N2O3.ClH.H2O/c1-28-24-11-5-4-10-22(24)26-15-13-25(14-16-26)17-20(27)18-29-23-12-6-8-19-7-2-3-9-21(19)23;;/h2-12,20,27H,13-18H2,1H3;1H;1H2

InChI key

INIDYCYXDNFBSV-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

α1-Adrenoceptor antagonist; antihypertensive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuo Tsuzaka et al.
International journal of urology : official journal of the Japanese Urological Association, 18(11), 792-795 (2011-09-16)
The aim of the present study was to compare the efficacy of the selective α(1D) -adrenoceptor antagonist naftopidil and the selective α(1A) -adrenoceptor antagonist silodosin (as an example) in the management of ureteral stones in Japanese male patients. A total
Hideyasu Tsumura et al.
International journal of radiation oncology, biology, physics, 81(4), e385-e392 (2011-06-15)
To compare the efficacy of three α(1A)/α(1D)-adrenoceptor (AR) antagonists--naftopidil, tamsulosin, and silodosin--that have differing affinities for the α(1)-AR subtypes in treating urinary morbidities in Japanese men with (125)I prostate implantation (PI) for prostate cancer. This single-institution prospective randomized controlled trial
G Sponer et al.
Journal of cardiovascular pharmacology, 20(6), 1006-1013 (1992-12-01)
The interaction of naftopidil with adrenoceptors was studied in comparison to standard drugs. Naftopidil binds specifically to alpha 1-adrenoceptors. The Ki values are 58.3 nM for naftopidil, 0.43 nM for prazosin, and 197 nM for urapidil. The affinities of naftopidil
S Yamada et al.
Life sciences, 50(2), 127-135 (1992-01-01)
Binding properties of naftopidil and alpha 1-adrenoceptor antagonists to alpha-adrenoceptors in prostates from benign prostatic hypertrophy (BPH) were characterized by radioreceptor assays using [3H]prazosin and [3H]rauwolscine. Specific binding of [3H]prazosin and [3H]rauwolscine in human prostatic membranes was saturable and of
Xiawen Liu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 907, 140-145 (2012-10-03)
Two bioanalytical HPLC methods (chiral solid phases (CSPs) HPLC and pre-column derivatization HPLC) were developed and validated for the determination of naftopidil enantiomers in rat plasma. Analytes were extracted from biomaterials by liquid-liquid extraction. The pre-column derivatization HPLC method employed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service