Key Documents
MTCP5200100U
MTC Bio ProPette LE Adjustable Pipettor
capacity 10-100 μL
Synonym(s):
pipette
About This Item
UNSPSC Code:
41121500
NACRES:
NB.15
Pricing and availability is not currently available.
feature
adjustable
manual
no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio P5200-100U
capacity
10-100 μL
General description
ProPette LE Single-Channel Pipettes provide maximum precision at virtually every conceivable volume. They feature a digital micrometer that is adjustable for precise volume control as well as a mechanical volume lock positioned at the top of the pipette that can be engaged or disengaged with one hand. These pipettors are lightweight and finely balanced for maximum ergonomics.
ProPette LE, 10 to 100µL
Features and Benefits
  • Adjustable with Roto-Lock
  • Light weight with reduced forces
  • Full volume range (0.1µL-10mL)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)