Skip to Content
Merck

Skip To

MTCC2000AST

MTC™ Bio SureSeal S™ Microcentrifuge Tube

capacity 1.5 mL, sterile, assorted colors, pack of 500 ea

Synonym(s):

centrifuge tube, plastic tube, MTC Bio C2000-AST, sample collection tube, sample receptacle, sample tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 ea
Please contact Customer Service for Availability
€23.50

About This Item

NACRES:
NB.15
UNSPSC Code:
41121703
Capacity:
1.5 mL
Material:
assorted colors , conical , polypropylene
Sterility:
sterile
Packaging:
pack of 500 ea
Parameter:
28,000 × g speed

€23.50


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

assorted colors , conical , polypropylene

sterility

sterile

packaging

pack of 500 ea

manufacturer/tradename

MTC™ Bio C2000-AST

parameter

28,000 × g speed

capacity

1.5 mL

General description

Designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation, and general sample/reagent storage. SureSeal(TM) S tubes are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28,000×g) and the usable temperature range (-90°C to +121°C). The full-sized tube cap (with frosted marking surface), combined with the unique ring and groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps.
Microtube w/ cap, 1.5ml, assorted colors, (B, R, G, O, P, Y), sterile, w/ self-standing bag & Stop-Pops™, 500/pk

Features and Benefits

  • Sterile comes standard – by electron beam radiation
  • Self-Standing bag – with gusseted bottom
  • Stop-Pops™ supplied – 10 cap locks in every bag

Legal Information

MTC is a trademark of MTC Bio
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Stop-Pops is a trademark of MTC Bio
SureSeal S is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library