Skip to Content
Merck
All Photos(1)

Key Documents

MBD0064

Sigma-Aldrich

ExoPolyzyme

For enzymatic lysis of extracellular biofilm matrix

Synonym(s):

Biofilm degrading enzyme mix, Extracellular matrix degrading enzymatic mixture

Sign Into View Organizational & Contract Pricing

Select a Size

MBD0064-1VL
€351.00

€351.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
MBD0064-1VL
€351.00

About This Item

UNSPSC Code:
42312305
NACRES:
NA.32

€351.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

packaging

pkg of 1 vial

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Biofilms are complex structures, which adhere to surfaces and comprise of one or more types of bacteria and fungi. They form an extracellular matrix (ECM) which is generally composed of secreted polymers such as exopolysaccharides (EPS), extracellular DNA (eDNA), peptides and lipids. ExoPolyzyme is a mixture of seven enzymes that break down different components residing in ECM, such as EPS’s and lipids. Each of these enzymes has been previously used to inhibit or remove biofilm. The original development was in collaboration with Scott Tighe and the Extreme Microbiome Project.

Application

ExoPolyzme can be used in biofilm research and as a gentler method for removing the EPS and lipids from the ECM in metagenomic protocols, providing better access to the bacterial cells within the biofilm and eDNA.

Features and Benefits

  • Mixture of seven enzymes that break down different components residing in ECM, such as EPS’s and lipids
  • Gently removes the EPS and lipids from the ECM in metagenomic protocols
  • Allows better access to the bacterial cells within the biofilm and eDNA

Components

The components in ExoPolyzyme (MBD0064) are:




  • α-Amylase
  • Cellulase
  • β-Glucosidase
  • Lyticase
  • Alginate lyase
  • Lipase
  • Hemicellulase

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service