Skip to Content
Merck
All Photos(1)

Key Documents

MAK556

Sigma-Aldrich

Hypochlorite (Hypochlorous Acid) Assay Kit

sufficient for 200 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

MAK556-1KT
€356.00

€356.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
MAK556-1KT
€356.00

About This Item

UNSPSC Code:
41116104
NACRES:
NA.84

€356.00


Please contact Customer Service for Availability

Request a Bulk Order

application(s)

environmental

detection method

colorimetric

relevant disease(s)

cancer; genitourinary disorders

storage temp.

−20°C

General description

Hypochlorite anion (ClO-) and its protonated form, hypochlorous acid (HClO) are critical reactive oxygen species (ROS) in biological systems. Uncontrolled production of hypochlorite (hypochlorous acid) can lead to tissue damage and diseases including arthritis, renal failure and cancers. In addition, sodium hypochlorite (NaClO) has been widely used as a bleaching agent for surface cleaning, odor removal and water disinfection in our daily lives. Exposure to large amount of sodium hypochlorite can lead to poisoning with the symptoms of serious breathing problems, stomach irritation, redness and pain on skin and eye. ​

The Hypochlorite (Hypochlorous Acid) Assay Kit offers a sensitive absorption-based assay for measuring hypochlorite (hypochlorous acid) with high specificity. Upon selective reaction with hypochlorite (hypochlorous) the colorless Hypochlorite Sensor generates a strong color product. The color signal can be measured by an absorption microplate reader at ~580 nm. With this assay kit, trace amounts of hypochlorite can be detected.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service