Skip to Content
Merck
All Photos(1)

Key Documents

MAK536

Sigma-Aldrich

Cytotoxicity Assay Kit

sufficient for 100 assays (bioluminescent)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€334.00

€334.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€334.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€334.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

pharmaceutical

detection method

bioluminescent

relevant disease(s)

cancer

shipped in

wet ice

storage temp.

−20°C

General description

Adenosine 5′-triphosphate (ATP) is the chemical energy for cellular metabolism and is often referred to as “energy currency" of the cell. ATP is produced only in living cells during photosynthesis and cellular respiration and consumed in cellular processes including biosynthetic reactions, motility and cell division. It is a key indicator of cellular activity and has been utilized as a measure of cell viability and cytotoxicity in research and drug discovery.

Features and Benefits

  • As low as 50 cells can be quantified.
  • Compatible with high-throughput handling systems.
  • No wash and reagent transfer steps are involved.

Suitability

The kit is suitable for:
  • Cell proliferation: effects of cytokines, growth factor, nutrients.
  • Cytotoxicity and apoptosis: evaluation of toxic compounds, anti-cancer antibodies, toxins, environmental pollutants etc.
  • Drug discovery: high-throughput screening for anticancer drugs.

Principle

The Cytotoxicity Assay Kit provides a rapid method to measure intracellular ATP, cell viability, and cytotoxicity. The single working reagent lyses cells to release ATP, which, in the presence of luciferase, immediately reacts with the substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATP concentration and hence number of living cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service