Skip to Content
Merck
All Photos(1)

Key Documents

MAK509

Sigma-Aldrich

Ferric Reducing Antioxidant Power (FRAP) Assay Kit

sufficient for 250 colorimetric tests

Synonym(s):

FRAP Colorimetric Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€394.00

€394.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€394.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€394.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

cosmetics
food and beverages
pharmaceutical

detection method

colorimetric

relevant disease(s)

Alzheimer′s disease; Parkinson′s disease; arthritis; diabetes; rheumatological diseases; neurodegenerative disorders; neurological disorders; endocrinological disorders

shipped in

ambient

storage temp.

2-8°C

General description

Ferric Reducing Antioxidant Power (FRAP) is a measure of antioxidant capacity quantified by the antioxidant′s potential to reduce ferric iron (III) to ferrous iron (II). Antioxidants protect cells from damage by reactive oxygen species that are produced by oxidation reactions. As oxidative stress contributes to the development of many diseases including Alzheimer′s disease, Parkinson′s disease, diabetes, rheumatoid arthritis and neurodegeneration, the use of antioxidants in pharmacology is intensively studied. Antioxidants are also widely used as dietary supplements and as preservatives in a wide range of products such as food, cosmetics, rubber and gasoline.

Features and Benefits

  • Linear detection range: 0.5 to 180 µM Fe3+ reduction potential
  • Compatible with high-throughput handling systems.
  • The optimized formulation has greatly enhanced reagent and signal stability.

Suitability

The kit is suitable for the determination of ferric reduction antioxidant potential in plant extracts, foods, vitamins, supplements, and biological samples such as serum, plasma, and urine.

Principle

The Ferric Reducing Antioxidant Power (FRAP) Assay Kit measures antioxidant potential in which Fe3+ is reduced by antioxidant to Fe2+. The resulting Fe2+ specifically forms a colored complex with a chromogen. The color intensity at 590 nm is proportional to FRAP in the sample.

Storage Class Code

10 - Combustible liquids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service