Skip to Content
Merck
All Photos(1)

Key Documents

MAK472

Sigma-Aldrich

Iron Assay Kit

sufficient for 250 colorimetric tests

Synonym(s):

Assay Kit for Iron Detection

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€436.00

€436.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€436.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€436.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

food and beverages
pharmaceutical

detection method

colorimetric

relevant disease(s)

anemia; hepatitis; gastrointestinal diseases; hematological disorder

shipped in

ambient

storage temp.

2-8°C

General description

Iron level in blood is a reliable diagnostic indicator of various disease states. Increased levels of iron concentration in blood are associated with red blood cell destruction, decreased blood cell survival, acute hepatitis, certain sideroblastic anemias, ingestion of iron-rich diets, defects in iron storage (e.g., pernicious anemia). Decreased levels of blood iron may result from insufficient iron ingestion from diets, chronic blood loss pathologies, or increased demand on iron storage as during normal pregnancy.

Features and Benefits

  • Linear detection range 27 µg/dL (4.8 µM) to 1,000 µg/dL (179 µM) iron in 96-well plate assay.
  • Compatible with high-throughput handling systems.
  • The optimized formulation has greatly enhanced reagent and signal stability.
  • Low interference in biological samples.
  • No pretreatments are needed. Assays can be directly performed on serum samples

Suitability

The kit is suitable for the detection of iron in biological samples such as serum and soil extracts, and for studying the effects of drugs on iron metabolism.

Principle

The Iron Assay Kit is designed to measure total iron directly in serum without pretreatment. The improved method utilizes a chromogen that forms a blue colored complex specifically with Fe2+. Fe3+ in the sample is reduced to Fe2+, allowing the assay to determine total iron concentration. The intensity of the color, measured at 590 nm, is directly proportional to the iron concentration in the sample.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service