Skip to Content
Merck
All Photos(1)

Key Documents

MAK468

Sigma-Aldrich

NAD/NADH Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€506.00

€506.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€506.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€506.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

detection method

colorimetric

relevant disease(s)

cancer

shipped in

wet ice

storage temp.

−20°C

General description

Pyridine nucleotides play an important role in metabolism, so there is interest in monitoring their concentration levels in metabolic systems. Quantitative determination of NAD+/NADH has applications in research pertaining to energy transformation and the redox state of cells or tissue. Nicotinamide adenine dinucleotide (NAD) is an enzymatic cofactor that plays a pivotal role in numerous redox reactions. Serving as an electron carrier, NAD seamlessly transitions between its oxidized form (NAD) and reduced form (NADH). Apart from its involvement in redox reactions, NAD assumes essential roles in ADP-ribosylation reactions and serves as a substrate for sirtuins.

Features and Benefits

  • The linear detection range of the kit is 0.05 - 10 micromolar (µM).
  • Compatible with high-throughput handling systems.
  • Simple procedure involves adding a single working reagent, and reading the optical density at time zero and 15 minutes at room temperature.

Suitability

The kit is suitable for NAD+/NADH concentration and ratio determination in cell or tissue extracts.

Principle

The NAD/NADH Assay Kit is based on a lactate dehydrogenase cycling reaction, in which the formed NADH reduces a formazan (MTT) reagent. The intensity of the reduced product color, measured at 565 nm, is proportional to the NAD+/NADH concentration in the sample. This assay is highly specific for NAD+/NADH with minimal interference (<1%) by NADP+/NADPH. The assay is a convenient method to measure NAD, NADH, and their ratio.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service