Skip to Content
Merck
All Photos(3)

Key Documents

MAK433

Sigma-Aldrich

Adenosine Quantification Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

Adenosine Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€721.00

€721.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€721.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€721.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

usage

sufficient for 100 fluorometric tests

input

plasma

detection method

fluorometric

relevant disease(s)

cancer; neurological disorders; cardiovascular diseases

shipped in

dry ice

storage temp.

−20°C

General description

Adenosine is a purine nucleoside base, which is the precursor of adenosine triphosphate, or ATP, and is utilized thoroughly throughout the entire body in general metabolism.[1] Extracellular adenosine acts as a local modulator with a generally cytoprotective function in the body.[2] Its effects on tissue protection and repair fall into four categories: increasing the ratio of oxygen supply to demand; protecting against ischaemic damage by cell conditioning; triggering anti-inflammatory responses; and the promotion of angiogenesis.[3]

Application

The Adenosine Quantification Assay Kit may be used for:
  • Cancer Research
  • Cardiovascular Disease Research
  • Neurological Disease Research

Features and Benefits

  • Supportive calculator (Click here to download a calculator excel file) Analyze results based on your experimental data!
  • Quick instruction bench card - to assure your experimental success
  • More experiments in one kit - contains sufficient reagents for 100 tests

Suitability

The kit is suitable for the quantification of adenosine in plasma.

Principle

The Adenosine Quantification Assay Kit provides a simple and quick fluorometric method for the detection of adenosine. The kit is based on a multi-step enzymatic approach, resulting in the formation of fluorescent product that is measured at λEx = 550 nm/λEm = 585. The kit has a linear range of 0 to 20 μM.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenosine receptors as therapeutic targets.
Jacobson K A, et al.
Nature Reviews. Drug Discovery, 5(3), 247-264 (2006)
B B Fredholm et al.
Pharmacological reviews, 53(4), 527-552 (2001-12-06)
Four adenosine receptors have been cloned and characterized from several mammalian species. The receptors are named adenosine A(1), A(2A), A(2B), and A(3). The A(2A) and A(2B) receptors preferably interact with members of the G(s) family of G proteins and the
Monika Samsel et al.
Postepy higieny i medycyny doswiadczalnej (Online), 67, 1189-1203 (2014-01-01)
Adenosine is an endogenous purine nucleoside that plays an important role in many biochemical processes. It acts through the four types of adenosine receptors: A1, A2A, A2B, A3 which are located i.a. in the immune, nervous, circulatory, respiratory or urinary

Questions

1–4 of 4 Questions  
  1. Is it possible to use MAK433 with media samples instead of plasma? The product is specified for human plasma, but is it suitable for conditioned cell culture media?

    1 answer
    1. The protocol is solely validated for use with plasma samples. Regrettably, there is no available data regarding its use with culture cell medium or other sample types. Experimental validation would be necessary. Additionally, considering that the kit has a linear range of 0.5 to 20 μM, it may be essential to optimize the required sample amount. However, it's important to note that guarantee can't be given for its performance.

      Helpful?

  2. For the Adenosine Quantification Assay Kit (Cat# MAK433), does this also detect ATP/ADP or just adenosine?

    1 answer
    1. MAK433-1KT Adenosine Quantification Assay Kit is specific for Adenosine. It does not measure ATP or ADP.

      Helpful?

  3. Is that possible to measure adenosine on cell culture's supernatant?

    1 answer
    1. This kit is intended for use with plasma. The end-user would have to determine suitability for use with cell culture supernatants. The lowest concentration used to generate the standard curve is 0.5 uM per well. Samples would have to have sufficient adenosine present for detection. See the link below to review the protocol:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/254/126/mak433bul.pdf

      Helpful?

  4. 혹시 이 제품이 abcam사에서 판매하는 Adenosine Assay Kit (Fluorometric) (ab211094) 제품과 동일하거나 비슷한 원리의 제품인지 알 수 있을까요?

    1 answer
    1. Both the "Adenosine Quantification Assay Kit" (Product number: MAK433) and the "Adenosine Assay Kit (Fluorometric)" (ab211094) from Abcam are assay kits designed to measure adenosine levels using a multi-enzymatic method.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service