Skip to Content
Merck
All Photos(1)

Key Documents

MAK390

Sigma-Aldrich

Alpha Galactosidase (α-Gal) Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Synonym(s):

Alpha-Gal Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

detection method

fluorometric

storage temp.

−20°C

General description

Alpha-Galactosidase (α-Gal) hydrolyzes alpha-galactosyl moieties found in glycolipids and glycoproteins. In mammals, a-Gal hydrolyzes poly- and oligosaccharides commonly found in dietary sources that are difficult to digest. Therefore, α-Gal is used in dietary supplements that help to reduce the production of intestinal gases due to consumption of certain foods. It is known total α-Gal activity is due to two major isozymes with unique, yet different thermostability profiles. Alpha-Galactosidase A, is thermolabile and represents approximately 90% of total α-Gal activity. Fabry Disease, a lysosomal disease disorder, is characterized by mutations in alpha-Galactosidase A. These mutations cause abnormal accumulation of glycosphingolipids in lysosomes.

Application


  • Exploration of mechanism of total triterpenoids from fruits of Chaenomeles speciosa against senescent GES-1 cells induced by D-galactose based on Gln/GLS1/a-KG metabolic axis and mitochondrial apoptosis signaling pathway.: This study investigates the effects of total triterpenoids on senescent GES-1 cells, exploring mechanisms involving the Gln/GLS1/a-KG metabolic axis and mitochondrial apoptosis signaling pathway (He et al., 2024).

  • Diagnosis of Fabry Disease Using Alpha-Galactosidase A Activity or LysoGb3 in Blood Fails to Identify Up to Two Thirds of Female Patients.: This research highlights the limitations of using alpha-galactosidase A activity and LysoGb3 in blood for diagnosing Fabry Disease, especially in female patients (Duro et al., 2024).

  • Pegunigalsidase alfa: a novel, pegylated recombinant alpha-galactosidase enzyme for the treatment of Fabry disease.: This review focuses on Pegunigalsidase alfa, a new pegylated recombinant enzyme for treating Fabry Disease, discussing its efficacy and potential benefits (Germain & Linhart, 2024).

Features and Benefits

The Alpha Galactosidase (α-Gal) Activity Assay Kit:
  • Detection limit: < 0.1μU.
  • Specific: does not detect beta galactosidase activity.

Suitability

The kit is suitable for the detection of α-Galactosidase activity in tissue homogenates (kidney, etc.), cell lysates (U937, etc.) and biological fluids (saliva, etc.).

Principle

The Alpha Galactosidase (α-Gal) Activity Assay Kit provides a simple, rapid way to monitor total α-Gal activity in a wide variety of biological samples. In this kit, α-Gal cleaves a synthetic specific substrate releasing a fluorophore, which can be easily quantified (λex = 360 nm/λem = 445 nm). The assay is specific (beta galactosidase activity is not detected), sensitive and can detect as low as 0.1 μU of α-Galactosidase activity.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service