Skip to Content
Merck
All Photos(2)

Key Documents

MAK343

Sigma-Aldrich

PEG Virus Precipitation Kit

sufficient for 200 preparations

Synonym(s):

Virus Concentration Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 200 preparations

application(s)

sample preservation

storage temp.

−20°C

Suitability

Suitable to concentrate retroviruses, baculoviruses, lentiviruses, and phages etc. in cell culture medium or environmental samples.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service