Skip to Content
Merck
All Photos(2)

Key Documents

MAK211

Sigma-Aldrich

Lipolysis Colorimetric Assay Kit

Sufficient for 100 Colorimetric tests

Synonym(s):

Lipolysis Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes

storage temp.

−20°C

General description

Lipolysis is the process of hydrolyzing triglycerides to free fatty acids and glycerol.[1] This process involves the action of adipose TG lipase (ATGL), hormone-sensitive lipase (HSL), and monoglyceride lipase.[2] Lipolysis maintains the energy balance during fasting and exercise by providing a substrate for oxidative metabolism. Lipolysis is regulated by nutritional factors and hormones. Problems with the regulation of lipolysis is associated with obesity, diabetes, and metabolic syndromes.[2]

Suitability

Suitable for the measurement of lipolysis in 3T3-L1 and adipocytes.

Principle

The Lipolysis (3T3-L1) Colorimetric Assay Kit contains a synthetic catecholamine (isoproterenol) that activates β-adrenergic receptors. This activates adenylate cyclase, converting ATP to cAMP. cAMP then activates the hydrolysis of triglycerides by hormone-sensitive lipase. Lipolysis is determined by measuring a colorimetric product with absorbance at 570 nm (A570) proportional to the amount of glycerol present.

Kit Components Only

Product No.
Description

  • Lipolysis Assay Buffer

  • Lipolysis Wash Buffer

  • Glycerol Assay Buffer

  • Glycerol Probe, in DMSO

  • Glycerol Enzyme Mix

  • Glycerol Standard, 100 mM

  • Isoproterenol, 10 mM

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipolysis?a highly regulated multi-enzyme complex mediates the catabolism of cellular fat stores.
Lass A, et al.
Progress in Lipid Research, 50(1), 14-27 (2011)
Control of fatty acid and glycerol release in adipose tissue lipolysis.
Langin D.
Comptes rendus biologies, 329(8), 598-607 (2006)
John M Dean et al.
Cell reports, 33(1), 108228-108228 (2020-10-08)
The Mediator complex relays regulatory signals from gene-specific transcription factors to the basal transcriptional machinery. However, the role of individual Mediator subunits in different tissues remains unclear. Here, we demonstrate that MED19 is essential for adipogenesis and maintenance of white

Questions

  1. What solvent is used for MAK211G (Isoproterenol 10 mM)?

    1 answer
    1. Isoproterenol is prepared in dH20.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service