Skip to Content
Merck
All Photos(2)

Documents

MAK032

Sigma-Aldrich

Zinc Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

cancer; genitourinary disorders; immunological diseases; neurological disorders

storage temp.

2-8°C

General description

Zinc, one of the most abundant trace metals in living organisms, contributes to a variety of biological functions including neurological development, nucleic acid metabolism, and signal transduction. Zinc is utilized as a cofactor for many enzymes, such as carbonic anhydrase, which is important to carbon dioxide regulation and carboxypeptidase, which cleaves peptide linkages. It is involved in various cellular processes, such as immune function, wound healing, protein synthesis, DNA synthesis and cell division. Zinc facilitates normal growth and development during pregnancy, childhood and adolescence. It also exhibits antioxidant properties and is essential for proper sense of taste and smell.
Zinc, one of the most abundant trace metals in living organisms, contributes to a variety of biological functions including neurological development, nucleic acid metabolism, and signal transduction. Zinc is utilized as a cofactor for many enzymes, such as carbonic anhydrase, which is important to carbon dioxide regulation, and carboxypeptidase, which cleaves peptide linkages.

The Zinc Assay kit provides a simple and direct procedure for measuring zinc in a variety of samples, including serum, plasma, urine, or cerebral spinal fluid (CSF). Zinc ions bind to a ligand, which results in a colorimetric (560 nm) product proportional to the amount of zinc present.

Suitability

Suitable for measuring zinc concentrations in a variety of samples, including serum, plasma, urine, or cerebral spinal fluid (CSF).

Principle

The Zinc Assay kit provides a simple and direct procedure for measuring zinc in a variety of samples, including serum, plasma, urine, or cerebral spinal fluid (CSF). Zinc ions bind to a ligand, which results in a colorimetric (560 nm) product proportional to the amount of zinc present.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Zinc: the metal of life.
Kaur K, et al.
Comprehensive Reviews in Food Science and Food Safety, 13(4), 358-376 (2014)
Daniela Grădinaru et al.
Experimental and therapeutic medicine, 22(6), 1393-1393 (2021-10-16)
Zinc deficiencies have been reported in numerous pathologies, such as diabetes mellitus, but also in the physiological process of ageing. Similarly, the end products of glycoxidation processes, advanced glycation end products (AGEs), are damaging compounds, a myriad of reports linking
Youmin Pu et al.
Annals of translational medicine, 10(5), 249-249 (2022-04-12)
Renal ischemic reperfusion injury (RIRI) is the most hackneyed cause of acute renal injury with high incidence. As a slit diaphragm (SD), TRPC6 (transient receptor potential channel 6) can maintain the structure and function of glomerular podocytes, and its activation
Copper and zinc, biological role and significance of copper/zinc imbalance.
Osredkar J and Sustar N
Journal of Clinical Toxicology, 3, 2161-0495 (2011)
T O Ajiboye et al.
International journal of antimicrobial agents, 53(4), 467-473 (2018-12-12)
Zinc is a highly coveted redox-inactive micronutrient required for the growth and virulence of Acinetobacter baumannii. In this study, the role of the zinc uptake regulator Zur in the susceptibility and oxidative stress response of A. baumannii to antibiotics was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service