Skip to Content
Merck
All Photos(2)

Key Documents

MAK013

Sigma-Aldrich

Glucose and Sucrose Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Glucose and Sucrose Colorimetric/Fluorometric Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes

storage temp.

−20°C

General description

Sucrose is a disaccharide that is hydrolyzed to glucose and fructose by invertase.[1] Glucose is the primary carbohydrate utilized for energy during cellular respiration.[2] Upon cellular uptake, glucose can be shunted towards breakdown via glycolysis or polymerized for energy storage.[2]

Application

Glucose and sucrose assay kit has been used to:
  • determine sucrose,[3] aerobic glycolysis and pyruvate kinase activity[4]
  • determine protein and sugar in strawberries[5]
  • determine plasma insulin and glucose levels in mice[6]
  • analyze plasma glucose and plasma lactate[7] and to quantify glucose/sucrose.[8]

Suitability

Suitable for use with various biological samples including serum, plasma, other body fluids, food, and growth media.

Principle

In this assay kit, glucose is oxidized via glucose oxidase resulting in a colorimetric (570 nm)/ fluorometric (λex = 535 nm/λem = 587 nm) product, proportional to the glucose present. To measure sucrose, invertase is added to the reaction to convert the sucrose to glucose and fructose. The free glucose can be subtracted from the total glucose to give the concentration of sucrose present. This kit has a linear range of detection between 0.2-1.0 nmole of glucose for the fluorometric assay and 2-10 nmoles of glucose for the colorimetric assay.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nuno F Silva-Soares et al.
BMC ecology, 17(1), 21-21 (2017-06-09)
Understanding how species adapt to new niches is a central issue in evolutionary ecology. Nutrition is vital for the survival of all organisms and impacts species fitness and distribution. While most Drosophila species exploit rotting plant parts, some species have
Swee-Suak Ko et al.
International journal of molecular sciences, 21(21) (2020-11-01)
Vanilla orchid, which is well-known for its flavor and fragrance, is cultivated in tropical and subtropical regions. This shade-loving plant is very sensitive to high irradiance. In this study, we show that vanilla chloroplasts started to have avoidance movement when
Biochemical Parameters in Toxicological Studies in Africa: Significance, Principle of Methods, Data Interpretation, and Use in Plant Screenings
Victor K.
Toxicological Survey of African Medicinal Plants (2014)
Cristiane Matavelli et al.
Journal of insect physiology, 82, 66-74 (2015-09-12)
Species coexist using the same nutritional resource by partitioning it either in space or time, but few studies explore how species-specific nutritional requirements allow partitioning. Zaprionus indianus and Drosophila simulans co-exist in figs by invading the fruit at different stages;
Tinghui Li et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 158, 401-409 (2020-10-31)
Bydureon® (Bdn) is a once-weekly injectable long-acting release (LAR) product for adults with type 2 diabetes based on PLGA microspheres encapsulating the glucagon like peptide (GLP-1) analog, exenatide. Despite its widespread use in type 2 diabetes treatment, little information has

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service