Skip to Content
Merck
All Photos(1)

Documents

M8258

Sigma-Aldrich

N-Methylindoxyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352103

storage temp.

2-8°C

SMILES string

CC(=O)Oc1cn(C)c2ccccc12

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Masson et al.
Biochimica et biophysica acta, 1597(2), 229-243 (2002-06-05)
Hydrolysis of the neutral substrate N-methylindoxyl acetate (NMIA) by wild-type human butyrylcholinesterase (BuChE) and peripheral site mutants (D70G, Y332A, D70G/Y332A) was found to follow the Michaelis-Menten kinetics. K(m) was 0.14 mM for wild-type, and 0.07-0.16 mM for D70G, Y332A and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service