Skip to Content
Merck
All Photos(1)

Key Documents

M7695

Sigma-Aldrich

Anti-Munc-18-3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MUNC18C, Anti-PSP, Anti-Syntaxin Binding Protein 3 (STXBP3), Anti-UNC-18C

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€533.00

€533.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€533.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€533.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~66 kDa

species reactivity

rat, mouse, human

technique(s)

western blot: 0.75-1.5 μg/mL using mouse brain extract (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Mammalian uncoordinated-18-3 (Munc-18-3) (also known as Munc-18c, syntaxin-binding protein 3, STXBP3) is ubiquitously expressed.

Immunogen

synthetic peptide corresponding to amino acids 578-592 located at the C-terminus of mouse Munc-18-3, conjugated to KLH. This sequence is highly conserved (~90% identity) in human and rat Munc-18-3.

Application

Anti-Munc-18-3 antibody produced in rabbit has been used in immunohistochemistry and western blotting[1]

Biochem/physiol Actions

Mammalian uncoordinated-18-3 (Munc-18-3) expressed in 3T3-L1 adipocytes has been shown to interact with syntaxin 2 and 4 and to inhibit the binding of syntaxin-4 to vesicle associated membrane protein 2 (VAMP2). Munc-18c has been suggested to play a role in the docking/fusion of glucose transporter type 4 (GLUT4)-containing vesicles with the cell membrane of 3T3-L1 adipocytes. Heterozygous knock-out of the Munc-18c gene in mice impairs insulin-stimulated GLUT4 translocation in skeletal muscle and increases the susceptibility for severe glucose intolerance.
Munc-18-3 has a role in glucose metabolism and is involved in controlling the translocation of glucose transporter type 4 (GLUT4) in adipocytes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lourdes Garrido-Sanchez et al.
PloS one, 8(5), e63937-e63937 (2013-05-24)
Munc18c is associated with glucose metabolism and could play a relevant role in obesity. However, little is known about the regulation of Munc18c expression. We analyzed Munc18c gene expression in human visceral (VAT) and subcutaneous (SAT) adipose tissue and its
Protein expression of PKCZ (Protein Kinase C Zeta), Munc18c, and Syntaxin-4 in the insulin pathway in endometria of patients with polycystic ovary syndrome (PCOS
<BIG>Rivero R, et al</BIG>
Reproductive Biology and Endocrinology, 10, 17-17 (2012)
Munc18c heterozygous knockout mice display increased susceptibility for severe glucose intolerance
Oh E, et al.
Diabetes, 54(3), 638-647 (2005)
Rodrigo Rivero et al.
Reproductive biology and endocrinology : RB&E, 10, 17-17 (2012-03-07)
Polycystic Ovary Syndrome (PCOS) is an endocrine-metabolic disorder commonly associated with insulin resistance (IR). Previous studies indicate about the expression of molecules involved in the insulin pathway in endometria of women with PCOS-IR. Therefore, the aim of the present study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service