Skip to Content
Merck
All Photos(2)

Key Documents

M7070

Sigma-Aldrich

Anti-MO25 antibody produced in rabbit

enhanced validation

~2 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CAB39, Anti-CGI-66, Anti-Calcium binding protein 39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~2 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using HEK-293T cells expressing human MO25 and PC12 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAB39(51719)
mouse ... Cab39(12283)
rat ... Cab39(301574)

General description

MO25 (mouse protein 25) is also termed calcium binding protein 39, CAB39 and CGI-66 is an Armadillo repeat domain scaffolding protein.
MO25 is a 40 kDa scaffolding protein that interacts with STRADα and LKB1 to regulate tumor suppression. MO25 activates LKB1 and also localizes LKB1 in the cytoplasm .

Specificity

Anti-MO25 antibody is specific for MO25 in humans, rats and mice. In immunoblotting, staining of the MO25 band is specifically inhibited by the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 308-326 located near the C-terminus of human MO25. This sequence is identical in mouse, rat, dog, and bovine MO25.

Application

Anti-MO25 antibody is suitable for use in western blot (0.5-1 μg/mL using HEK-293T cells expressing human MO25 and PC12 cell lysates).
Anti-MO25 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

MO25 (mouse protein 25) binds to Liver kinase B1 (LKB1) and STE20-related kinase adaptor (STRAD) and activates LKB1 and anchors it in the cell cytosol. LKB1 is a key regulator of AMP activated protein kinase (AMPK), which is a master regulator of cellular energy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of the LKB1-STRAD-MO25 complex
Boudeau J, et al.
Journal of Cell Science, 117(26), 6365-6375 (2004)
LKB1-dependent signaling pathways
Alessi DR, et al.
Annual Review of Biochemistry, 75, 137-163 (2006)
A new mechanism for LKB1 activation
Lee SW and Lin HK
Molecular & Cellular Oncology, 5(3), e1035691-e1035691 (2018)
Marked phenotypic differences of endurance performance and exercise-induced oxygen consumption between AMPK and LKB1 deficiency in mouse skeletal muscle: changes occurring in the diaphragm
Miura S, et al.
American Journal of Physiology. Endocrinology and Metabolism, 305(2), E213-E229 (2013)
Marked phenotypic differences of endurance
Bruce CR, et al.
American Journal of Physiology. Endocrinology and Metabolism, 305, E213-E229 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service