Skip to Content
Merck
All Photos(2)

Key Documents

M5890

Sigma-Aldrich

Methyl 1,2,3,4-tetra-O-acetyl-β-D-glucuronate

≥98% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O11
CAS Number:
Molecular Weight:
376.31
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥98% (GC)

form

powder

optical activity

[α]20/D 8.1 to 9.5 °, c = 1% (w/v) in chloroform

technique(s)

gas chromatography (GC): suitable

storage temp.

2-8°C

SMILES string

COC(=O)C1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

InChI

1S/C15H20O11/c1-6(16)22-10-11(23-7(2)17)13(24-8(3)18)15(25-9(4)19)26-12(10)14(20)21-5/h10-13,15H,1-5H3

InChI key

DPOQCELSZBSZGX-UHFFFAOYSA-N

Application

Methyl 1,2,3,4-tetra-O-acetyl-β-D-glucuronate has been used in a study to assess design and synthesis of a water-soluble glucuronide derivatives of camptothecin for cancer prodrug monotherapy and antibody-directed enzyme prodrug therapy. [1] It has also been used in a study to investigate the synthesis and role of glycosylthio heterocycles in carbohydrate chemistry. [2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y L Leu et al.
Journal of medicinal chemistry, 42(18), 3623-3628 (1999-09-10)
Glucuronide prodrugs of 9-aminocamptothecin were synthesized. Prodrug 4, in which 9-aminocamptothecin was connected to glucuronic acid by an aromatic spacer via a carbamate linkage, was stable in both aqueous solution and human plasma. Prodrug 4 and its potassium salt 12
Synthesis and role of glycosylthio heterocycles in carbohydrate chemistry
Ashry, E., et al.
Tetrahedron, 62, 2943-2998 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service