Skip to Content
Merck
All Photos(2)

Documents

M5421

Sigma-Aldrich

Magnesium hydroxide

BioXtra, ≥95%

Synonym(s):

Magnesium dihydroxide, Mg(OH)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(OH)2
CAS Number:
Molecular Weight:
58.32
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

product line

BioXtra

Assay

≥95%

impurities

≤0.01% Phosphorus (P)
≤0.1% Insoluble matter

mp

350 °C (lit.)

solubility

5 M HCl: 0.1 M, clear, yellow

anion traces

chloride (Cl-): ≤0.5%

cation traces

Al: ≤0.02%
Ca: ≤0.7%
Cu: ≤0.0005%
Fe: ≤0.02%
K: ≤0.005%
NH4+: ≤0.05%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.002%

SMILES string

O[Mg]O

InChI

1S/Mg.2H2O/h;2*1H2/q+2;;/p-2

InChI key

VTHJTEIRLNZDEV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium hydroxide has been used in the synthesis of brucite.

Biochem/physiol Actions

Magnesium hydroxide is a flame retardant. Magnesium hydroxide is commonly used as an antacid which has a long-lasting effect. The liquid formulation of magnesium hydroxide is called milk of magnesia. It is used as a laxative. It is used in the treatment of diarrhea along with aluminium hydroxide. The naturally occurring form of magnesium hydroxide is brucite.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fire Retardancy of Polymeric Materials, 293-293 (2000)
Kinetics of the chrysotile and brucite dehydroxylation reaction: a combined non-isothermal/isothermal thermogravimetric analysis and high-temperature X-ray powder diffraction study
Trittschack R, et al.
Physics and Chemistry of Minerals, 41(3), 197-214 (2014)
Plastics Additives: A Rapra Market Report, 57-57 (2005)
Lehne's Pharmacotherapeutics for Advanced Practice Providers - E-Book, 714-714 (2017)
Suresh Kumar Kailasa et al.
The Analyst, 137(19), 4490-4496 (2012-08-11)
We report a dispersive liquid-liquid microextraction (LLME) method using surface modified Mg(OH)(2) nanoparticles (NPs) with oleic acid (OA) as hydrophobic affinity probes for the extraction and preconcentration of hydrophobic biomolecules prior to MALDI MS analysis. OA-capped Mg(OH)(2) NPs were characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service