Skip to Content
Merck
All Photos(1)

Key Documents

M4656

Sigma-Aldrich

(±)-p-Methoxyamphetamine hydrochloride

Synonym(s):

PMA hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO · HCl
CAS Number:
Molecular Weight:
201.69
EC Number:
MDL number:
UNSPSC Code:
12352200

drug control

Home Office Schedule 1; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(CC(C)N)cc1

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul D Callaghan et al.
Journal of neurochemistry, 100(3), 617-627 (2006-12-22)
p-Methoxyamphetamine (PMA) has been implicated in fatalities as a result of 'ecstasy' (MDMA) overdose worldwide. Like MDMA, acute effects are associated with marked changes in serotonergic neurotransmission, but the long-term effects of PMA are poorly understood. The aim of this
M M W Straiko et al.
Neuroscience, 144(1), 223-231 (2006-11-07)
The present study quantified the cleaved form of the microtubule-associated protein tau (cleaved MAP-tau, C-tau), a previously demonstrated marker of CNS toxicity, following the administration of monoamine-depleting regimens of the psychostimulant drugs amphetamine (AMPH), methamphetamine (METH), +/-3,4-methylenedioxymethamphetamine (MDMA), or para-methoxyamphetamine
Amparo Fornés et al.
The Biochemical journal, 412(3), 495-506 (2008-03-18)
The neuronal glycine transporter GLYT2 controls the availability of the neurotransmitter in glycinergic synapses, and the modulation of its function may influence synaptic transmission. The active transporter is located in membrane rafts and reaches the cell surface through intracellular trafficking.
Fatal paramethoxy-amphetamine (PMA) poisoning in the Australian Capital Territory.
Paul G Lamberth et al.
The Medical journal of Australia, 188(7), 426-426 (2008-04-09)
Diego Bustamante et al.
Pharmacology, biochemistry, and behavior, 79(2), 199-212 (2004-10-27)
The analgesic effects of (+)- and (-)-amphetamine (AMPH), (+/-)-p-methoxyamphetamine (MA), (+/-)-N-methyl-p-methoxyamphetamine (MMA) and (+/-)-N-ethyl-p-methoxyamphetamine (EMA) were compared using two different algesimetric tests in rats. In the formalin test, (+)-AMPH elicited significant antinociception at doses of 0.2, 2 and 8 mg/kg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service