Skip to Content
Merck
All Photos(1)

Key Documents

M4017

Sigma-Aldrich

N-Methyl-L-glutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO4
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CN[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C6H11NO4/c1-7-4(6(10)11)2-3-5(8)9/h4,7H,2-3H2,1H3,(H,8,9)(H,10,11)/t4-/m0/s1

InChI key

XLBVNMSMFQMKEY-BYPYZUCNSA-N

Biochem/physiol Actions

N-Methyl-L-glutamic acid is an amino acid derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Goto et al.
Journal of the American Chemical Society, 131(14), 5040-5041 (2009-03-24)
In the prokaryotic translation system formylmethionyl-tRNA(fMet)(CAU) acts as an exclusive initiator to yield peptides bearing formylmethionine at the N-terminus. We herein report a new methodology to initiate the translation reaction with peptidyl-tRNA(fMet)(CAU) in which the peptidyl group consists of unusual
Kristin R McKenna et al.
The Analyst, 145(24), 8008-8015 (2020-10-15)
Carbohydrates are the most abundant class of biomolecules on Earth with a diverse array of biological functions. It is hypothesized that they likely had an important role in the development of life on the primoridal Earth as well. Since sugars

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service