Skip to Content
Merck
All Photos(1)

Key Documents

M2017

Sigma-Aldrich

Mazindol

≥98% (TLC), powder

Synonym(s):

5-(4-Chlorophenyl)-2,5-dihydro-3H-imidazo[2,1-a]isoindol-5-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClN2O
CAS Number:
Molecular Weight:
284.74
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

color

white

solubility

DMSO: 10 mg/mL

originator

Wyeth

SMILES string

OC1(C2=CC=C(Cl)C=C2)C3=CC=CC=C3C4=NCCN14

InChI

1S/C16H13ClN2O/c17-12-7-5-11(6-8-12)16(20)14-4-2-1-3-13(14)15-18-9-10-19(15)16/h1-8,20H,9-10H2

InChI key

ZPXSCAKFGYXMGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anorectic agent; inhibits the dopamine, norepinephrine and serotonin transporters with Kis of 27.6 nM, 3.2 nM, and 153 nM, respectively.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Inoue
Obesity research, 3 Suppl 4, 549S-552S (1995-11-01)
An anoerxiant, mazindol suppresses food intake by 1) stimulating beta-adrenergic receptors, 2) inhibiting the feeding center and, 3) stimulating the satiety center in the hypothalamus. In Japan, mazindol is available for clinical use. We examined the effects of mazindol on
Kasper Severinsen et al.
Molecular pharmacology, 85(2), 208-217 (2013-11-12)
Mazindol has been explored as a possible agent in cocaine addiction pharmacotherapy. The tetracyclic compound inhibits both the dopamine transporter and the serotonin transporter, and simple chemical modifications considerably alter target selectivity. Mazindol, therefore, is an attractive scaffold for both
W Michael Caudle et al.
Neurotoxicology, 26(4), 721-728 (2005-08-23)
Although banned in the 1970s, significant levels of the organochlorine pesticide heptachlor are still present in the environment raising concern over potential human exposure. In particular, organochlorine pesticides have been linked to an increased risk of Parkinson's disease. Studies from
E L Barker et al.
The Journal of biological chemistry, 273(31), 19459-19468 (1998-07-25)
Human and Drosophila melanogaster serotonin (5-HT) transporters (SERTs) exhibit similar 5-HT transport kinetics and can be distinguished pharmacologically by many, but not all, biogenic amine transporter antagonists. By using human and Drosophila SERT chimeras, major determinants of potencies of two
Johannes Backs et al.
Cardiovascular research, 67(2), 283-290 (2005-07-12)
Endothelin-1 (ET-1) potentiates norepinephrine (NE)-induced contractile responses. An impairment of cardiac NE re-uptake by the neuronal NE transporter (NET) contributes to an increased NE net release in failing hearts. We hypothesized that both phenomena are caused by ET-1-mediated inhibition of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service