Skip to Content
Merck
All Photos(1)

Key Documents

M1630

Sigma-Aldrich

O-Methyl-L-threonine

≥98.0% (TLC)

Synonym(s):

L-Threonine methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO3
CAS Number:
Molecular Weight:
133.15
Beilstein:
1722343
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Product Name

O-Methyl-L-threonine,

Assay

≥98.0% (TLC)

form

powder

color

white to off-white

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CO[C@H](C)[C@H](N)C(O)=O

InChI

1S/C5H11NO3/c1-3(9-2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t3-,4+/m1/s1

InChI key

FYCWLJLGIAUCCL-DMTCNVIQSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitors of specific aminoacyl-tRNA synthetases prevent thyrotropin-induced desensitization in cultured human thyroid cells.
S Filetti et al.
The Journal of biological chemistry, 257(3), 1342-1346 (1982-02-10)
P P Dembure et al.
Blood, 57(6), 1125-1131 (1981-06-01)
Hemoglobin beta-chain synthesis by rabbit erythroid cells was tested for dependence on availability of complementary alpha-chains. Reticulocytes and bone marrow cells were obtained from variant rabbits that have hemoglobin with isoleucine in alpha-chains but not in beta-chains. This characteristic permits
Daniel Osorio-Gómez et al.
Learning & memory (Cold Spring Harbor, N.Y.), 24(1), 14-23 (2016-12-17)
The insular cortex (IC) is required for conditioned taste aversion (CTA) retrieval. However, it remains unknown which cortical neurotransmitters levels are modified upon CTA retrieval. Using in vivo microdialysis, we observed that there were clear elevations in extracellular glutamate, norepinephrine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service