Skip to Content
Merck
All Photos(1)

Key Documents

M1272

Sigma-Aldrich

Anti-Mouse IgA (α-chain specific) antibody produced in sheep

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€193.00

€193.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€193.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

€193.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

Ouchterlony double diffusion: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

General description

IgA antibody is secretory antibody and is present abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears and saliva. Thus secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces
Anti-Mouse IgA (α-chain specific) is specific for α chain of mouse IgA.

Immunogen

Purified mouse IgA (myeloma protein)

Application

Anti-Mouse IgA (α-chain specific) is suitable for Ouchterlony Double Diffusion. The antibody was used to detect antigens to Helicobacter hepaticus in mice serum using a concentration of 5 μg/ml.

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2

Reconstitution

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark T Whary et al.
Immunology, 133(2), 165-178 (2011-03-24)
To model inflammatory bowel disease, we assessed infection with Helicobacter hepaticus 3B1 (ATCC 51449) and a potential probiotic Lactobacillus reuteri (ATCC PTA-6475) in gnotobiotic B6.129P2-IL-10(tm1Cgn) (IL-10(-/-) ) mice. No typhlocolitis developed in germ-free controls (n=21) or in L. reuteri (n=8)
Jan Holmgren et al.
Nature medicine, 11(4 Suppl), S45-S53 (2005-04-07)
There is currently great interest in developing mucosal vaccines against a variety of microbial pathogens. Mucosally induced tolerance also seems to be a promising form of immunomodulation for treating certain autoimmune diseases and allergies. Here we review the properties of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service